siRNA Details
| Gene Symbol: TCEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080390.3 | NP_525129.1 | 3 | 359 | ||
| GenBank mRNA | AF325115.1 | - | 3 | 304 | ||
| AK312138.1 | - | 3 | 332 | |||
| BC093932.1 | - | 2 | 237 | 7939777 | ||
| BC104764.1 | - | 2 | 238 | 8143767 | ||
| CD580311.2 | - | 3 | 390 | |||

TCEAL2
Genomic Map