siRNA Details
| Gene Symbol: AQP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080429.2 | NP_536354.2 | 4 | 468 | ||
| XM_011510104.2 | XP_011508406.1 | 4 | 528 | |||
| GenBank mRNA | AB066105.1 | - | 4 | 468 | ||
| AF159174.1 | - | 4 | 464 | |||
| BC069607.1 | - | 4 | 468 | 7262209 | ||
| BC074896.2 | - | 4 | 465 | 30915249 | ||
| BC074897.2 | - | 4 | 465 | 30915478 | ||

AQP10
Genomic Map