siRNA Details
| Gene Symbol: C11orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080659.2 | NP_542390.2 | 4 | 252 | ||
| GenBank mRNA | AK057948.1 | - | 4 | 190 | ||
| BC008501.1 | - | 4 | 216 | 4294167 | ||
| BQ102454.1 | - | 4 | 252 | |||
| Gene Symbol: HSPB2-C11orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037651.1 | 5 | 639 | |||
| GenBank mRNA | BC110872.1 | - | 5 | 634 | 6193385 | |

C11orf52
Genomic Map