siRNA Details
| Gene Symbol: GLYATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001220494.2 | NP_001207423.1 | 5, 6 | 606 | ||
| NM_001220496.2 | NP_001207425.1 | 4, 5 | 434 | |||
| NM_080661.4 | NP_542392.2 | 4, 5 | 337 | |||
| XM_011545354.2 | XP_011543656.1 | 4, 5 | 453 | |||
| XM_011545355.2 | XP_011543657.1 | 4, 5 | 439 | |||
| XM_011545356.2 | XP_011543658.1 | 5, 6 | 498 | |||
| XM_011545357.2 | XP_011543659.1 | 5, 6 | 484 | |||
| XM_011545358.2 | XP_011543660.1 | 5, 6 | 1022 | |||
| XM_011545359.2 | XP_011543661.1 | 4, 5 | 978 | |||
| GenBank mRNA | AB207212.1 | - | 2, 3 | 194 | ||
| AF087878.1 | - | 4, 5 | 469 | |||
| AK055818.1 | - | 3, 4 | 591 | |||
| AK091965.1 | - | 4, 5 | 337 | |||
| AK130017.1 | - | 1, 2 | 1191 | |||
| BC008353.1 | - | 5, 6 | 534 | 3537224 | ||
| DC339353.1 | - | 4, 5 | 561 | |||
| DQ084381.1 | - | 4, 5 | 434 | |||

GLYATL1
Genomic Map