siRNA Details
| Gene Symbol: WDR89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008726.2 | NP_001008726.1 | 4 | 550 | ||
| NM_001258272.1 | NP_001245201.1 | 3 | 946 | |||
| NM_080666.3 | NP_542397.1 | 3 | 441 | |||
| XM_011536383.2 | XP_011534685.1 | 2 | 480 | |||
| XM_011536385.2 | XP_011534687.1 | 4 | 448 | |||
| GenBank mRNA | AF115513.1 | - | 3 | 373 | ||
| AK095324.1 | - | 3 | 389 | |||
| BC010698.1 | - | 2 | 333 | 3870073 | ||
| BI603622.1 | - | 3 | 397 | |||
| BP278550.1 | - | 4 | 461 | |||
| BX248008.1 | - | 3 | 430 | |||
| DA882425.1 | - | 3 | 442 | |||

WDR89
Genomic Map