siRNA Details
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042681.1 | NP_001036146.1 | 23 | 7556 | ||
| NM_001042682.1 | NP_001036147.1 | 13 | 5855 | |||
| NM_012102.3 | NP_036234.3 | 24 | 7741 | |||
| XM_005263464.2 | XP_005263521.1 | 23 | 7495 | |||
| XM_005263466.1 | XP_005263523.1 | 17 | 6378 | |||
| XM_011541510.1 | XP_011539812.1 | 22 | 7481 | |||
| XM_017001358.1 | XP_016856847.1 | 23 | 7246 | |||
| XM_017001359.1 | XP_016856848.1 | 23 | 7751 | |||
| GenBank mRNA | AB007927.2 | - | 22 | 6890 | ||
| AB036737.1 | - | 24 | 7566 | |||
| AK074834.1 | - | 1 | 2114 | |||
| AL050176.1 | - | 1 | 1551 | |||

RERE
Genomic Map