siRNA Details
| Gene Symbol: EHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206615.1 | NP_001193544.1 | 7 | 803 | ||
| NM_001206616.1 | NP_001193545.1 | 8 | 873 | |||
| NM_012153.5 | NP_036285.2 | 8 | 872 | |||
| XM_005252857.1 | XP_005252914.1 | 8 | 870 | |||
| XM_005252860.1 | XP_005252917.1 | 8 | 726 | |||
| XM_005252861.1 | XP_005252918.1 | 7 | 804 | |||
| XM_005252862.1 | XP_005252919.1 | 7 | 673 | |||
| XM_011519983.1 | XP_011518285.1 | 8 | 898 | |||
| XM_011519984.1 | XP_011518286.1 | 9 | 978 | |||
| XM_011519985.1 | XP_011518287.1 | 8 | 763 | |||
| XM_017017534.1 | XP_016873023.1 | 7 | 801 | |||
| GenBank mRNA | AF124438.1 | - | 7 | 709 | ||
| AF124439.1 | - | 8 | 778 | |||
| AF170583.1 | - | 8 | 782 | |||
| AF203977.1 | - | 8 | 793 | |||
| AF212848.1 | - | 8 | 812 | |||
| AK309318.1 | - | 8 | 898 | |||
| AK310867.1 | - | 8 | 873 | |||
| AY882601.1 | - | 7 | 681 | |||
| BC038995.2 | - | 8 | 832 | 6055934 | ||

EHF
Genomic Map