siRNA Details
| Gene Symbol: ZSWIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080603.4 | NP_542170.3 | 2 | 1795 | ||
| GenBank mRNA | BC001672.2 | - | 2 | 1763 | 2988536 | |
| BC080525.1 | - | 2 | 1763 | 4547006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSWIM1| Gene Symbol: ZSWIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080603.4 | NP_542170.3 | 2 | 1795 | ||
| GenBank mRNA | BC001672.2 | - | 2 | 1763 | 2988536 | |
| BC080525.1 | - | 2 | 1763 | 4547006 | ||