siRNA Details
| Gene Symbol: CBLN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080617.5 | NP_542184.1 | 3 | 1853 | ||
| GenBank mRNA | AK289640.1 | - | 3 | 1657 | ||
| AY358527.1 | - | 3 | 888 | |||
| BC050026.1 | - | 3 | 1855 | 4820943 | ||
| BC069402.1 | - | 3 | 552 | 7262170 | ||
| BC069488.1 | - | 3 | 552 | 7262182 | ||
| BC069516.1 | - | 3 | 552 | 7262099 | ||

CBLN4
Genomic Map