siRNA Details
| Gene Symbol: SLC35A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080670.3 | NP_542401.1 | 3 | 2246 | ||
| GenBank mRNA | AF445025.1 | - | 2 | 3499 | ||
| AJ420598.1 | - | 1 | 1259 | |||
| AK122601.1 | - | 4 | 1308 | |||
| AL833972.1 | - | 2 | 1965 | |||
| BC009906.2 | - | 3 | 1976 | 2961449 | ||

SLC35A4
Genomic Map