siRNA Details
| Gene Symbol: TMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080751.2 | NP_542789.2 | 3 | 253 | ||
| XM_005260660.3 | XP_005260717.1 | 1 | 447 | |||
| GenBank mRNA | AF417580.2 | - | 3 | 253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMC2| Gene Symbol: TMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080751.2 | NP_542789.2 | 3 | 253 | ||
| XM_005260660.3 | XP_005260717.1 | 1 | 447 | |||
| GenBank mRNA | AF417580.2 | - | 3 | 253 | ||