siRNA Details
| Gene Symbol: WFDC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080753.2 | NP_542791.1 | 1, 2 | 159 | ||
| GenBank mRNA | AY038182.1 | - | 1, 2 | 159 | ||
| BC131579.1 | - | 1, 2 | 125 | 40112102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC10A| Gene Symbol: WFDC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080753.2 | NP_542791.1 | 1, 2 | 159 | ||
| GenBank mRNA | AY038182.1 | - | 1, 2 | 159 | ||
| BC131579.1 | - | 1, 2 | 125 | 40112102 | ||