siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145350.1 | NP_001138822.1 | 3 | 500 | ||
| NR_026933.2 | 3 | 500 | ||||
| XM_006723710.2 | XP_006723773.1 | 3 | 500 | |||
| XM_011528577.1 | XP_011526879.1 | 3 | 500 | |||
| GenBank mRNA | AK058089.1 | - | 2 | 475 | ||
| AK310337.1 | - | 3 | 500 | |||
| BC112060.1 | - | 1 | 220 | 8327528 | ||
| BC112062.1 | - | 1 | 220 | 8327530 | ||
| DB057743.1 | - | 3 | 500 | |||

C20orf173
Genomic Map