siRNA Details
| Gene Symbol: DPCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080870.3 | NP_543146.2 | 2 | 3603 | ||
| GenBank mRNA | AB064272.1 | - | 1 | 131 | ||
| AL833738.1 | - | 1 | 945 | |||
| BC069477.1 | - | 1 | 129 | 7262259 | ||
| BC101661.1 | - | 1 | 194 | 8069167 | ||
| BC101663.1 | - | 1 | 194 | 8069169 | ||

DPCR1
Genomic Map