siRNA Details
| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080927.3 | NP_563615.3 | 1, 2 | 576 | ||
| GenBank mRNA | AB073146.1 | - | 1, 2 | 576 | ||
| AF387547.1 | - | 1, 2 | 224 | |||
| AK056350.1 | - | 1, 2 | 569 | |||
| AK302649.1 | - | 1, 2 | 56 | |||
| BC007117.1 | - | 2, 3 | 123 | |||
| BC029658.1 | - | 1, 2 | 95 | 4431275 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028523, BC066097, AF387548.

DCBLD2
Genomic Map