siRNA Details
| Gene Symbol: SLC39A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280557.1 | NP_001267486.1 | 2 | 94 | ||
| NM_017767.2 | NP_060237.2 | 9 | 1739 | |||
| NM_130849.3 | NP_570901.2 | 10 | 1595 | |||
| GenBank mRNA | AK000334.1 | - | 9 | 1719 | ||
| AK000489.1 | - | 2 | 89 | |||
| AK000695.1 | - | 9 | 1654 | |||
| AK025537.1 | - | 10 | 1589 | |||
| AK056900.1 | - | 2 | 88 | |||
| AK290563.1 | - | 10 | 1543 | |||
| BC001688.2 | - | 2 | 66 | |||
| BC062625.1 | - | 10 | 1575 | 5203858 | ||
| BE798833.1 | - | 1 | 225 | |||
| KU178707.1 | - | 10 | 1489 | |||
| KU178708.1 | - | 9 | 1207 | |||

SLC39A4
Genomic Map