siRNA Details
Gene Symbol: TRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282902.2 | NP_001269831.1 | 6 | 5182 | ||
NM_001282903.2 | NP_001269832.1 | 7 | 5671 | |||
NM_014112.4 | NP_054831.2 | 7 | 5685 | |||
XM_005251049.2 | XP_005251106.1 | 6 | 5500 | |||
XM_006716625.1 | XP_006716688.1 | 7 | 5622 | |||
XM_011517264.1 | XP_011515566.1 | 8 | 5755 | |||
XM_011517266.2 | XP_011515568.1 | 7 | 5490 | |||
XM_011517267.1 | XP_011515569.1 | 7 | 5608 | |||
XM_011517268.1 | XP_011515570.1 | 7 | 5597 | |||
XM_017013801.1 | XP_016869290.1 | 6 | 9284 | |||
GenBank mRNA | AF183810.1 | - | 7 | 5679 | ||
AF264784.1 | - | 6 | 5228 | |||
AK021470.1 | - | 1 | 374 | |||
BC125020.1 | - | 7 | 5515 | 40073132 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
