siRNA Details
| Gene Symbol: TMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080751.2 | NP_542789.2 | 3 | 370 | ||
| XM_005260660.3 | XP_005260717.1 | 1 | 564 | |||
| GenBank mRNA | AF417580.2 | - | 3 | 370 | ||
| AK294013.1 | - | 1 | 116 | |||
| AK309782.1 | - | 1 | 126 | |||
| AK316261.1 | - | 1 | 83 | |||
| DA769512.1 | - | 1 | 83 | |||

TMC2
Genomic Map