siRNA Details
| Gene Symbol: ASB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080868.2 | NP_543144.1 | 1 | 315 | ||
| NR_026546.1 | 1 | 315 | ||||
| GenBank mRNA | AF403035.1 | - | 1 | 175 | ||
| AF429968.1 | - | 1 | 292 | |||
| AK098606.1 | - | 1 | 315 | |||
| BC022517.1 | - | 1 | 248 | |||
| BC036573.2 | - | 1 | 293 | 5297837 | ||

ASB17
Genomic Map