siRNA Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080878.2 | NP_543154.1 | 3, 4 | 256 | ||
| XM_011509210.1 | XP_011507512.1 | 5, 6 | 733 | |||
| XM_011509211.1 | XP_011507513.1 | 5, 6 | 730 | |||
| GenBank mRNA | AY065973.1 | - | 3, 4 | 228 | ||
| AY358905.1 | - | 3, 4 | 256 | |||
| BC117225.1 | - | 3, 4 | 254 | 40125776 | ||
| BC143341.1 | - | 3, 4 | 254 | 9051846 | ||

ITLN2
Genomic Map