siRNA Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142733.2 | NP_001136205.2 | 8 | 1313 | ||
| NM_130387.5 | NP_569058.1 | 1 | 337 | |||
| XM_017005736.1 | XP_016861225.1 | 8 | 1663 | |||
| XM_017005737.1 | XP_016861226.1 | 8 | 1627 | |||
| GenBank mRNA | AF403032.1 | - | 1 | 337 | ||
| AL832472.1 | - | 8 | 1314 | |||

ASB14
Genomic Map