siRNA Details
| Gene Symbol: TOR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134431.2 | NP_001127903.1 | 2 | 463 | ||
| NM_130459.3 | NP_569726.2 | 3 | 729 | |||
| GenBank mRNA | AK027677.1 | - | 3 | 664 | ||
| AK055034.1 | - | 2 | 408 | |||
| AK092388.1 | - | 1 | 1980 | |||
| BC100907.1 | - | 2 | 398 | |||
| BC100908.2 | - | 2 | 398 | |||
| BC100909.2 | - | 2 | 393 | |||
| DB168410.1 | - | 1 | 141 | |||

TOR2A
Genomic Map