siRNA Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305017.1 | NP_001291946.1 | 3 | 256 | ||
| NM_130897.2 | NP_570967.1 | 3 | 263 | |||
| NR_130942.1 | 3 | 804 | ||||
| NR_130943.1 | 4 | 353 | ||||
| XM_006721295.3 | XP_006721358.1 | 3 | 460 | |||
| XM_006721296.3 | XP_006721359.1 | 3 | 213 | |||
| XM_011523370.2 | XP_011521672.1 | 3 | 220 | |||
| GenBank mRNA | AF125108.1 | - | 3 | 258 | ||
| BC054892.1 | - | 4 | 284 | 6619254 | ||
| CD676360.1 | - | 3 | 256 | |||
| DB458006.1 | - | 3 | 225 | |||
| HY018370.1 | - | 3 | 264 | |||
| HY123054.1 | - | 3 | 246 | |||

DYNLRB2
Genomic Map