siRNA Details
| Gene Symbol: FAM71B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130899.2 | NP_570969.2 | 1 | 279 | ||
| GenBank mRNA | AF465821.1 | - | 1 | 255 | ||
| AK057543.1 | - | 1 | 275 | |||
| AY973323.1 | - | 1 | 179 | |||
| BC022035.1 | - | 1 | 287 | 4826350 | ||
| BC025397.1 | - | 1 | 280 | 4827541 | ||
| BC025409.1 | - | 1 | 280 | 4825198 | ||
| BC025998.1 | - | 1 | 280 | 4836822 | ||

FAM71B
Genomic Map