siRNA Details
| Gene Symbol: OTUD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130901.1 | NP_570971.1 | 2, 3 | 416 | ||
| GenBank mRNA | AJ430383.1 | - | 2, 3 | 416 | ||
| AK126002.1 | - | 7, 8 | 822 | |||
| AK308630.1 | - | 5, 6 | 617 | |||
| BC035668.1 | - | 4, 5 | 492 | |||

OTUD7A
Genomic Map