siRNA Details
| Gene Symbol: AMOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113490.1 | NP_001106962.1 | 2 | 1368 | ||
| NM_133265.2 | NP_573572.1 | 3 | 937 | |||
| XM_005262087.1 | XP_005262144.1 | 5 | 2113 | |||
| XM_005262090.1 | XP_005262147.1 | 2 | 610 | |||
| XM_011530875.2 | XP_011529177.1 | 6 | 2225 | |||
| XM_017029289.1 | XP_016884778.1 | 4 | 1141 | |||
| GenBank mRNA | AB028994.3 | - | 1 | 141 | ||
| AF286598.1 | - | 3 | 937 | |||
| AK301573.1 | - | 1604 | ||||
| AY987378.1 | - | 2 | 1368 | |||
| BC094712.1 | - | 3 | 1443 | |||
| BC130294.1 | - | 1 | 358 | 40146325 | ||

AMOT
Genomic Map