siRNA Details
| Gene Symbol: TMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080751.2 | NP_542789.2 | 10, 11 | 1247 | ||
| XM_005260660.3 | XP_005260717.1 | 8, 9 | 1441 | |||
| GenBank mRNA | AF417580.2 | - | 10, 11 | 1247 | ||
| AK294013.1 | - | 8, 9 | 993 | |||
| AK316261.1 | - | 8, 9 | 963 | |||

TMC2
Genomic Map