siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145350.1 | NP_001138822.1 | 6 | 1007 | ||
| XM_006723710.2 | XP_006723773.1 | 5 | 955 | |||
| GenBank mRNA | AK058089.1 | - | 5 | 982 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf173| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145350.1 | NP_001138822.1 | 6 | 1007 | ||
| XM_006723710.2 | XP_006723773.1 | 5 | 955 | |||
| GenBank mRNA | AK058089.1 | - | 5 | 982 | ||