siRNA Details
Gene Symbol: DPCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080870.3 | NP_543146.2 | 3 | 5222 | ||
GenBank mRNA | AB064272.1 | - | 2 | 1750 | ||
AL833738.1 | - | 2 | 2564 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080870.3 | NP_543146.2 | 3 | 5222 | ||
GenBank mRNA | AB064272.1 | - | 2 | 1750 | ||
AL833738.1 | - | 2 | 2564 |