siRNA Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080878.2 | NP_543154.1 | 8 | 1134 | ||
| XM_011509210.1 | XP_011507512.1 | 10 | 1611 | |||
| XM_011509211.1 | XP_011507513.1 | 10 | 1608 | |||
| GenBank mRNA | AY065973.1 | - | 8 | 1106 | ||
| AY358905.1 | - | 8 | 1134 | |||

ITLN2
Genomic Map