siRNA Details
| Gene Symbol: DBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004395.3 | NP_004386.2 | 7, 8 | 890 | ||
| NM_080881.2 | NP_543157.1 | 8, 9 | 1006 | |||
| XM_005265827.3 | XP_005265884.1 | 7, 8 | 837 | |||
| XM_006714826.2 | XP_006714889.1 | 7, 8 | 727 | |||
| XM_011534446.2 | XP_011532748.1 | 8, 9 | 1009 | |||
| XM_011534447.2 | XP_011532749.1 | 7, 8 | 1506 | |||
| XM_017009139.1 | XP_016864628.1 | 7, 8 | 1008 | |||
| XM_017009140.1 | XP_016864629.1 | 7, 8 | 999 | |||
| GenBank mRNA | AK094125.1 | - | 6, 7 | 936 | ||
| AK225523.1 | - | 7, 8 | 831 | |||
| AK314645.1 | - | 7, 8 | 831 | |||
| AL110225.1 | - | 8, 9 | 1335 | |||
| BC000283.1 | - | 7, 8 | 814 | 3356428 | ||
| BC007281.1 | - | 7, 8 | 814 | 3343626 | ||
| BC007567.1 | - | 7, 8 | 814 | 3028196 | ||
| D17530.1 | - | 7, 8 | 815 | |||
| U00802.1 | - | 7, 8 | 790 | |||

DBN1
Genomic Map