siRNA Details
| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080927.3 | NP_563615.3 | 16 | 3497 | ||
| XM_011512419.2 | XP_011510721.1 | 15 | 3213 | |||
| GenBank mRNA | AB073146.1 | - | 16 | 3497 | ||
| AF104313.1 | - | 1 | 42 | |||
| AK001362.1 | - | 1 | 499 | |||
| BC029658.1 | - | 16 | 3059 | 4431275 | ||

DCBLD2
Genomic Map