siRNA Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130388.3 | NP_569059.3 | 2 | 997 | ||
| GenBank mRNA | AF403030.1 | - | 1 | 777 | ||
| AK096896.1 | - | 2 | 977 | |||
| AK300684.1 | - | 3 | 877 | |||
| BC069436.1 | - | 1 | 777 | 7262244 | ||
| BC069555.1 | - | 1 | 777 | 7262256 | ||
| BC093883.1 | - | 1 | 838 | 7939728 | ||
| BC104946.1 | - | 1 | 838 | 8143949 | ||

ASB12
Genomic Map