siRNA Details
| Gene Symbol: GRIN3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133445.2 | NP_597702.2 | 2 | 1422 | ||
| XM_011518211.2 | XP_011516513.1 | 2 | 846 | |||
| GenBank mRNA | AB075853.1 | - | 2 | 1422 | ||
| AF416558.1 | - | 2 | 834 | |||
| AJ416950.2 | - | 2 | 836 | |||
| AK127342.1 | - | 2 | 389 | |||
| BC132866.1 | - | 2 | 1248 | 40146888 | ||
| BC132868.1 | - | 2 | 1248 | 40146890 | ||
| BC167432.1 | - | 2 | 1422 | 8860109 | ||

GRIN3A
Genomic Map