siRNA Details
| Gene Symbol: ZFP82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321917.1 | NP_001308846.1 | 3 | 353 | ||
| NM_001321918.1 | NP_001308847.1 | 3 | 353 | |||
| NM_001321919.1 | NP_001308848.1 | 3 | 353 | |||
| NM_133466.3 | NP_597723.1 | 3 | 353 | |||
| GenBank mRNA | AK074942.1 | - | 3 | 335 | ||
| AK315709.1 | - | 3 | 313 | |||
| AL834267.1 | - | 3 | 327 | |||
| BC032577.2 | - | 3 | 320 | 5492906 | ||
| BX491216.1 | - | 3 | 354 | |||
| GU388429.1 | - | 3 | 323 | |||

ZFP82
Genomic Map