siRNA Details
| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138294.2 | NP_612151.1 | 4 | 189 | ||
| GenBank mRNA | AF462605.1 | - | 4 | 185 | ||
| BC039863.1 | - | 4 | 189 | 5745088 | ||
| BC107044.2 | - | 3 | 149 | 40002653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PATE1| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138294.2 | NP_612151.1 | 4 | 189 | ||
| GenBank mRNA | AF462605.1 | - | 4 | 185 | ||
| BC039863.1 | - | 4 | 189 | 5745088 | ||
| BC107044.2 | - | 3 | 149 | 40002653 | ||