siRNA Details
| Gene Symbol: TMEM129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127266.1 | NP_001120738.1 | 2 | 1047 | ||
| NM_138385.3 | NP_612394.1 | 2 | 1047 | |||
| XM_005248038.4 | XP_005248095.1 | 2 | 919 | |||
| XM_005248039.4 | XP_005248096.1 | 2 | 488 | |||
| XM_011513594.2 | XP_011511896.1 | 2 | 497 | |||
| XM_017008834.1 | XP_016864323.1 | 2 | 784 | |||
| GenBank mRNA | BC009331.2 | - | 2 | 1047 | ||
| BC018921.2 | - | 2 | 1047 | |||
| BC126370.1 | - | 3 | 937 | 8992086 | ||

TMEM129
Genomic Map