siRNA Details
| Gene Symbol: REEP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138393.1 | NP_612402.1 | 5 | 1179 | ||
| GenBank mRNA | AK058112.1 | - | 6 | 1154 | ||
| AX775999.1 | - | 5 | 1179 | |||
| BC008201.1 | - | 5 | 1179 | 3865853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

REEP6| Gene Symbol: REEP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138393.1 | NP_612402.1 | 5 | 1179 | ||
| GenBank mRNA | AK058112.1 | - | 6 | 1154 | ||
| AX775999.1 | - | 5 | 1179 | |||
| BC008201.1 | - | 5 | 1179 | 3865853 | ||