siRNA Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138395.3 | NP_612404.1 | 1 | 609 | ||
| GenBank mRNA | AB107013.1 | - | 1 | 552 | ||
| AK098121.1 | - | 1 | 544 | |||
| BC009115.1 | - | 1 | 512 | |||
| BC126294.1 | - | 1 | 547 | 8992010 | ||

MARS2
Genomic Map