siRNA Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138409.2 | NP_612418.2 | 2 | 307 | ||
| XM_011535400.2 | XP_011533702.1 | 2 | 233 | |||
| XM_017010219.1 | XP_016865708.1 | 3 | 605 | |||
| XM_017010220.1 | XP_016865709.1 | 2 | 874 | |||
| XM_017010221.1 | XP_016865710.1 | 2 | 608 | |||
| GenBank mRNA | AK090775.1 | - | 2 | 307 | ||
| AK292736.1 | - | 2 | 238 | |||
| AK307078.1 | - | 2 | 236 | |||
| BC010003.2 | - | 2 | 252 | 4134852 | ||
| BC039855.1 | - | 2 | 170 | 3850270 | ||

MRAP2
Genomic Map