siRNA Details
| Gene Symbol: HELQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297755.1 | NP_001284684.1 | 2 | 742 | ||
| NM_001297756.1 | NP_001284685.1 | 2 | 742 | |||
| NM_001297757.1 | NP_001284686.1 | 2 | 742 | |||
| NM_001297758.1 | NP_001284687.1 | 2 | 631 | |||
| NM_001297759.1 | NP_001284688.1 | 2 | 742 | |||
| NM_133636.3 | NP_598375.2 | 2 | 742 | |||
| NR_123737.1 | 2 | 870 | ||||
| XM_005262711.1 | XP_005262768.1 | 2 | 603 | |||
| XM_005262713.2 | XP_005262770.1 | 2 | 832 | |||
| XM_006714076.2 | XP_006714139.1 | 2 | 704 | |||
| XM_011531580.2 | XP_011529882.1 | 2 | 704 | |||
| XM_017007679.1 | XP_016863168.1 | 2 | 603 | |||
| XM_017007680.1 | XP_016863169.1 | 2 | 832 | |||
| XM_017007681.1 | XP_016863170.1 | 2 | 704 | |||
| XM_017007682.1 | XP_016863171.1 | 2 | 810 | |||
| XM_017007683.1 | XP_016863172.1 | 1 | 130 | |||
| GenBank mRNA | AF436845.1 | - | 2 | 703 | ||
| AK126255.1 | - | 1 | 130 | |||
| AK128665.1 | - | 2 | 841 | |||
| AK308395.1 | - | 2 | 682 | |||
| AK312725.1 | - | 2 | 705 | |||
| BC015428.1 | - | 2 | 843 | |||
| BC095473.1 | - | 2 | 539 | |||
| BC111502.1 | - | 2 | 663 | |||
| KU178845.1 | - | 2 | 552 | |||
| KU178846.1 | - | 2 | 680 | |||

HELQ
Genomic Map