siRNA Details
| Gene Symbol: ZNF551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270938.1 | NP_001257867.1 | 3 | 566 | ||
| NM_138347.4 | NP_612356.2 | 3 | 688 | |||
| NR_073102.1 | 2 | 564 | ||||
| GenBank mRNA | AK091273.1 | - | 3 | 648 | ||
| AK300465.1 | - | 2 | 526 | |||
| AK307670.1 | - | 3 | 566 | |||
| BC005868.2 | - | 3 | 620 | 3530863 | ||
| BX538151.1 | - | 2 | 534 | |||

ZNF551
Genomic Map