siRNA Details
| Gene Symbol: SCRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145023.1 | NP_001138495.1 | 4 | 543 | ||
| NM_138355.3 | NP_612364.2 | 4 | 543 | |||
| XM_005257776.4 | XP_005257833.1 | 4 | 523 | |||
| XM_017025286.1 | XP_016880775.1 | 4 | 471 | |||
| GenBank mRNA | AK290646.1 | - | 4 | 446 | ||
| AK303835.1 | - | 4 | 499 | |||
| BC002980.1 | - | 4 | 486 | |||
| BC010408.2 | - | 4 | 457 | 3945715 | ||
| BC017317.2 | - | 4 | 456 | 3832079 | ||

SCRN2
Genomic Map