siRNA Details
| Gene Symbol: MCU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270680.1 | NP_001257609.1 | 5 | 908 | ||
| NM_138357.2 | NP_612366.1 | 5 | 631 | |||
| NR_073062.1 | 7 | 796 | ||||
| XM_017016882.1 | XP_016872371.1 | 5 | 831 | |||
| GenBank mRNA | AK075439.1 | - | 5 | 619 | ||
| AK128016.1 | - | 5 | 908 | |||
| AK301475.1 | - | 7 | 777 | |||
| AK310455.1 | - | 8 | 801 | |||
| AK311133.1 | - | 7 | 790 | |||
| AK315519.1 | - | 5 | 614 | |||
| BC034235.1 | - | 5 | 631 | 5296557 | ||

MCU
Genomic Map