siRNA Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138414.2 | NP_612423.1 | 7 | 730 | ||
| XM_017022894.1 | XP_016878383.1 | 7 | 743 | |||
| GenBank mRNA | AK057008.1 | - | 7 | 689 | ||
| BC011981.1 | - | 7 | 721 | 3863665 | ||
| Gene Symbol: LOC613038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW136592.1 | - | 3 | 459 | ||

SGF29
Genomic Map