siRNA Details
Gene Symbol: ZFP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321917.1 | NP_001308846.1 | 4 | 483 | ||
NM_001321918.1 | NP_001308847.1 | 4 | 480 | |||
NM_001321919.1 | NP_001308848.1 | 4 | 480 | |||
NM_133466.3 | NP_597723.1 | 4 | 480 | |||
GenBank mRNA | AK074942.1 | - | 4 | 462 | ||
AK315709.1 | - | 4 | 440 | |||
AL834267.1 | - | 4 | 454 | |||
BC032577.2 | - | 4 | 447 | 5492906 | ||
BX491216.1 | - | 4 | 481 | |||
GU388429.1 | - | 4 | 450 |
