siRNA Details
| Gene Symbol: C6orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025260.3 | NP_079536.2 | 5 | 649 | ||
| NM_138272.2 | NP_612116.1 | 5 | 630 | |||
| NM_138273.2 | NP_612117.1 | 4 | 558 | |||
| NM_138274.2 | NP_612118.1 | 3 | 517 | |||
| NM_138275.2 | NP_612119.1 | 3 | 498 | |||
| NM_138277.2 | NP_612121.1 | 5 | 649 | |||
| XM_011514920.2 | XP_011513222.1 | 5 | 664 | |||
| XM_011514921.2 | XP_011513223.1 | 5 | 683 | |||
| XM_017011330.1 | XP_016866819.1 | 3 | 710 | |||
| XM_017011331.1 | XP_016866820.1 | 3 | 948 | |||
| XM_017011332.1 | XP_016866821.1 | 4 | 592 | |||
| GenBank mRNA | AJ292259.1 | - | 5 | 609 | ||
| AJ292260.1 | - | 5 | 590 | |||
| AJ292261.1 | - | 4 | 518 | |||
| AJ292262.1 | - | 3 | 477 | |||
| AJ292263.1 | - | 3 | 458 | |||
| AJ292264.1 | - | 4 | 727 | |||
| AJ292265.1 | - | 5 | 609 | |||
| AK092392.1 | - | 5 | 620 | |||
| BC113719.1 | - | 5 | 649 | 8322771 | ||
| BC113721.1 | - | 5 | 649 | 8322773 | ||
| BC143850.1 | - | 5 | 630 | 9052366 | ||

C6orf25
Genomic Map