siRNA Details
Gene Symbol: PATE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138294.2 | NP_612151.1 | 5 | 701 | ||
GenBank mRNA | AF462605.1 | - | 5 | 697 | ||
BC039863.1 | - | 5 | 699 | 5745088 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PATE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138294.2 | NP_612151.1 | 5 | 701 | ||
GenBank mRNA | AF462605.1 | - | 5 | 697 | ||
BC039863.1 | - | 5 | 699 | 5745088 |