siRNA Details
| Gene Symbol: COPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138336.1 | NP_612209.1 | 5 | 729 | ||
| XM_017003411.1 | XP_016858900.1 | 5 | 1154 | |||
| GenBank mRNA | AF453951.1 | - | 5 | 729 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COPS9| Gene Symbol: COPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138336.1 | NP_612209.1 | 5 | 729 | ||
| XM_017003411.1 | XP_016858900.1 | 5 | 1154 | |||
| GenBank mRNA | AF453951.1 | - | 5 | 729 | ||