siRNA Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138348.4 | NP_612357.4 | 7 | 1849 | ||
| XM_011514154.2 | XP_011512456.1 | 6 | 5459 | |||
| XM_017010015.1 | XP_016865504.1 | 6 | 5652 | |||
| GenBank mRNA | AK092203.1 | - | 3 | 1169 | ||
| BC007706.2 | - | 5 | 1422 | 3952677 | ||
| CK817982.1 | - | 1 | 331 | |||

OTULIN
Genomic Map